siRNA Details
| Gene Symbol: DENND2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256404.1 | NP_001243333.1 | 5 | 1535 | ||
| NM_198459.3 | NP_940861.3 | 3 | 1166 | |||
| GenBank mRNA | AK308302.1 | - | 3 | 1187 | ||
| BX647950.1 | - | 3 | 1167 | |||
| BX649075.1 | - | 2 | 174 | |||
| CR749576.1 | - | 5 | 1459 | |||

DENND2C
Genomic Map