siRNA Details
| Gene Symbol: OLFML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198474.3 | NP_940876.2 | 2 | 669 | ||
| XM_005252878.4 | XP_005252935.1 | 3 | 446 | |||
| GenBank mRNA | AK298137.1 | - | 3 | 264 | ||
| AY358591.1 | - | 2 | 271 | |||
| BC035895.1 | - | 2 | 496 | 5229159 | ||
| BC092488.1 | - | 2 | 514 | 30531479 | ||
| BX647908.1 | - | 2 | 557 | |||
| DB257515.1 | - | 2 | 537 | |||

OLFML1
Genomic Map