siRNA Details
| Gene Symbol: TPRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198485.3 | NP_940887.1 | 6 | 2188 | ||
| XM_005247382.4 | XP_005247439.1 | 6 | 5432 | |||
| XM_011512732.2 | XP_011511034.2 | 7 | 4318 | |||
| XM_011512735.2 | XP_011511037.1 | 5 | 5256 | |||
| XM_017006257.1 | XP_016861746.1 | 14 | 3480 | |||
| XM_017006258.1 | XP_016861747.1 | 11 | 2908 | |||
| XM_017006259.1 | XP_016861748.1 | 13 | 4091 | |||
| XM_017006260.1 | XP_016861749.1 | 13 | 3287 | |||
| XM_017006261.1 | XP_016861750.1 | 10 | 3036 | |||
| XM_017006262.1 | XP_016861751.1 | 7 | 2326 | |||
| XM_017006263.1 | XP_016861752.1 | 5 | 1904 | |||
| GenBank mRNA | AK125682.1 | - | 6 | 2188 | ||

TPRG1
Genomic Map