siRNA Details
| Gene Symbol: CCDC84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198489.2 | NP_940891.1 | 3, 4 | 445 | ||
| NR_104049.1 | 3, 4 | 445 | ||||
| NR_104050.1 | 3, 4 | 445 | ||||
| GenBank mRNA | AB094093.1 | - | 3, 4 | 436 | ||
| BC126469.1 | - | 3, 4 | 410 | 8992185 | ||
| BC126471.1 | - | 3, 4 | 410 | 8992187 | ||
| BC144209.1 | - | 3, 4 | 410 | |||
| BC144210.1 | - | 3, 4 | 410 | |||
| HY073607.1 | - | 3, 4 | 447 | |||

CCDC84
Genomic Map