siRNA Details
| Gene Symbol: FAM92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198491.2 | NP_940893.1 | 9 | 1818 | ||
| XM_005255931.2 | XP_005255988.1 | 9 | 1153 | |||
| GenBank mRNA | AK126284.1 | - | 9 | 1818 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM92B| Gene Symbol: FAM92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198491.2 | NP_940893.1 | 9 | 1818 | ||
| XM_005255931.2 | XP_005255988.1 | 9 | 1153 | |||
| GenBank mRNA | AK126284.1 | - | 9 | 1818 | ||