siRNA Details
| Gene Symbol: LANCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170331.1 | NP_001163802.1 | 3 | 1018 | ||
| NM_198511.2 | NP_940913.1 | 3 | 1018 | |||
| XM_011543904.2 | XP_011542206.1 | 3 | 503 | |||
| GenBank mRNA | AK124915.1 | - | 3 | 1018 | ||
| BC093667.1 | - | 3 | 810 | 7939512 | ||
| BC093669.1 | - | 3 | 810 | 7939514 | ||
| BC143528.1 | - | 3 | 810 | |||

LANCL3
Genomic Map