siRNA Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198514.3 | NP_940916.2 | 10 | 2130 | ||
| XM_011539769.2 | XP_011538071.1 | 10 | 2130 | |||
| GenBank mRNA | AK090631.1 | - | 10 | 2101 | ||
| AK098487.1 | - | 8 | 1244 | |||
| AL832090.1 | - | 1 | 101 | |||
| BC032598.1 | - | 6 | 894 | 5529619 | ||
| BX538287.1 | - | 6 | 924 | |||
| BX647641.1 | - | 8 | 1501 | |||

NHLRC2
Genomic Map