siRNA Details
| Gene Symbol: HDDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286451.1 | NP_001273380.1 | 4 | 631 | ||
| NM_198527.3 | NP_940929.1 | 4 | 782 | |||
| NR_104447.1 | 4 | 674 | ||||
| GenBank mRNA | AK057584.1 | - | 1 | 1350 | ||
| BC033794.1 | - | 4 | 782 | 5122164 | ||
| CV573934.1 | - | 4 | 616 | |||

HDDC3
Genomic Map