siRNA Details
| Gene Symbol: DRAXIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198545.3 | NP_940947.3 | 4 | 855 | ||
| XM_011541405.2 | XP_011539707.1 | 5 | 1029 | |||
| XM_017001214.1 | XP_016856703.1 | 5 | 1165 | |||
| GenBank mRNA | AB301919.1 | - | 3 | 715 | ||
| AK075558.1 | - | 4 | 850 | |||
| AY358750.1 | - | 4 | 816 | |||
| BC021286.2 | - | 2 | 205 | |||
| BC111064.1 | - | 4 | 857 | 5769984 | ||

DRAXIN
Genomic Map