siRNA Details
| Gene Symbol: DRAXIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075558.1 | - | 8 | 1841 | ||
| AK092318.1 | - | 1 | 1779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DRAXIN| Gene Symbol: DRAXIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075558.1 | - | 8 | 1841 | ||
| AK092318.1 | - | 1 | 1779 | |||