siRNA Details
| Gene Symbol: MIGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270384.1 | NP_001257313.1 | 16 | 4522 | ||
| NM_198549.3 | NP_940951.1 | 16 | 4519 | |||
| XM_005270846.4 | XP_005270903.1 | 15 | 4429 | |||
| XM_005270847.4 | XP_005270904.1 | 16 | 5200 | |||
| XM_005270848.4 | XP_005270905.1 | 16 | 4809 | |||
| XM_011541422.2 | XP_011539724.1 | 16 | 4573 | |||
| XM_017001227.1 | XP_016856716.1 | 16 | 4489 | |||
| XM_017001228.1 | XP_016856717.1 | 16 | 4806 | |||
| XM_017001229.1 | XP_016856718.1 | 16 | 4581 | |||
| XM_017001230.1 | XP_016856719.1 | 16 | 4594 | |||
| GenBank mRNA | BX537792.1 | - | 16 | 4519 | ||

MIGA1
Genomic Map