siRNA Details
Gene Symbol: FAM89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198552.2 | NP_940954.1 | 2 | 1111 | ||
GenBank mRNA | BC009447.1 | - | 2 | 1102 | 3530481 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198552.2 | NP_940954.1 | 2 | 1111 | ||
GenBank mRNA | BC009447.1 | - | 2 | 1102 | 3530481 |