siRNA Details
| Gene Symbol: C3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198562.2 | NP_940964.1 | 3 | 1016 | ||
| GenBank mRNA | AK096799.1 | - | 2 | 761 | ||
| AK125642.1 | - | 3 | 960 | |||
| BC023586.2 | - | 3 | 1850 | 4650297 | ||
| BC053629.1 | - | 3 | 1016 | 6204001 | ||
| BC061697.1 | - | 3 | 1006 | 6737728 | ||

C3orf62
Genomic Map