siRNA Details
| Gene Symbol: NAT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198571.2 | NP_940973.2 | 4 | 2615 | ||
| XM_005250325.1 | XP_005250382.1 | 5 | 2594 | |||
| XM_011516168.1 | XP_011514470.1 | 4 | 2576 | |||
| GenBank mRNA | AK092119.1 | - | 3 | 1831 | ||
| AK096556.1 | - | 4 | 2615 | |||
| AL831911.1 | - | 2 | 1885 | |||

NAT16
Genomic Map