siRNA Details
| Gene Symbol: SPEM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097400.1 | - | 2 | 1372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPEM1| Gene Symbol: SPEM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097400.1 | - | 2 | 1372 | ||