siRNA Details
| Gene Symbol: CCDC62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201435.4 | NP_958843.2 | 4 | 575 | ||
| NR_027918.2 | 4 | 575 | ||||
| XM_006719643.1 | XP_006719706.1 | 4 | 483 | |||
| XM_006719644.1 | XP_006719707.1 | 4 | 483 | |||
| XM_011538856.1 | XP_011537158.1 | 4 | 483 | |||
| XM_011538857.1 | XP_011537159.1 | 4 | 483 | |||
| GenBank mRNA | AK097663.1 | - | 4 | 499 | ||
| AK292466.1 | - | 4 | 483 | |||
| AY254201.1 | - | 4 | 556 | |||
| BC060796.1 | - | 4 | 558 | 5284906 | ||

CCDC62
Genomic Map