siRNA Details
| Gene Symbol: CIAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004804.2 | NP_004795.1 | 7 | 2193 | ||
| GenBank mRNA | AK056423.1 | - | 5 | 2559 | ||
| AK092687.1 | - | 1 | 710 | |||
| AK124703.1 | - | 1 | 1796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIAO1| Gene Symbol: CIAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004804.2 | NP_004795.1 | 7 | 2193 | ||
| GenBank mRNA | AK056423.1 | - | 5 | 2559 | ||
| AK092687.1 | - | 1 | 710 | |||
| AK124703.1 | - | 1 | 1796 | |||