siRNA Details
| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006407.3 | NP_006398.1 | 3 | 807 | ||
| GenBank mRNA | AB052638.1 | - | 3 | 705 | ||
| AB097051.1 | - | 3 | 788 | |||
| AF070523.1 | - | 3 | 813 | |||
| AF115965.1 | - | 3 | 798 | |||
| AF125530.1 | - | 3 | 799 | |||
| AF161476.1 | - | 3 | 788 | |||
| AK074612.1 | - | 3 | 759 | |||
| AK223566.1 | - | 3 | 711 | |||
| AK301637.1 | - | 4 | 721 | |||
| AK303148.1 | - | 3 | 693 | |||
| BC005143.2 | - | 3 | 730 | 3510436 | ||
| BC020797.1 | - | 3 | 724 | 4134658 | ||

ARL6IP5
Genomic Map