siRNA Details
| Gene Symbol: SNRNP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006857.2 | NP_006848.1 | 4, 5 | 399 | ||
| NR_037862.1 | 4, 5 | 399 | ||||
| GenBank mRNA | AK291128.1 | - | 4, 5 | 380 | ||
| AK312041.1 | - | 4, 5 | 364 | |||
| BC017890.1 | - | 4, 5 | 384 | 4699198 | ||
| BU171817.1 | - | 4, 5 | 390 | |||
| X76302.1 | - | 4, 5 | 383 | |||

SNRNP27
Genomic Map