siRNA Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198595.1 | NP_001185524.1 | 5 | 4833 | ||
| NM_006873.3 | NP_006864.2 | 4 | 4716 | |||
| GenBank mRNA | AK023617.1 | - | 1 | 1585 | ||
| CR936807.1 | - | 5 | 4816 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STON1| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198595.1 | NP_001185524.1 | 5 | 4833 | ||
| NM_006873.3 | NP_006864.2 | 4 | 4716 | |||
| GenBank mRNA | AK023617.1 | - | 1 | 1585 | ||
| CR936807.1 | - | 5 | 4816 | |||