siRNA Details
| Gene Symbol: FAM107A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076778.2 | NP_001070246.1 | 4 | 3078 | ||
| NM_001282713.1 | NP_001269642.1 | 4 | 2878 | |||
| NM_001282714.1 | NP_001269643.1 | 5 | 3326 | |||
| NM_007177.3 | NP_009108.1 | 5 | 3176 | |||
| GenBank mRNA | AF089853.1 | - | 4 | 3169 | ||
| AF089854.1 | - | 5 | 3267 | |||

FAM107A
Genomic Map