siRNA Details
Gene Symbol: APPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012096.2 | NP_036228.1 | 2 | 282 | ||
XM_011533583.2 | XP_011531885.1 | 3 | 335 | |||
GenBank mRNA | AB037849.2 | - | 2 | 135 | ||
AF169797.1 | - | 2 | 193 | |||
AF424738.1 | - | 2 | 191 | |||
AK299008.1 | - | 3 | 324 | |||
AK310716.1 | - | 2 | 230 | |||
BC028599.2 | - | 2 | 282 | 4829430 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145221, AK088080, AK045438, BU514763, AK033566, and others.
Panther Classification:
Gene Ontology Categories:
