siRNA Details
| Gene Symbol: G3BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012297.4 | NP_036429.2 | 2 | 249 | ||
| NM_203504.2 | NP_987100.1 | 2 | 384 | |||
| NM_203505.2 | NP_987101.1 | 2 | 384 | |||
| XM_005263382.2 | XP_005263439.1 | 2 | 358 | |||
| XM_005263383.3 | XP_005263440.1 | 3 | 323 | |||
| XM_011532441.2 | XP_011530743.1 | 2 | 244 | |||
| XM_017008876.1 | XP_016864365.1 | 2 | 232 | |||
| XM_017008877.1 | XP_016864366.1 | 2 | 411 | |||
| XM_017008878.1 | XP_016864367.1 | 2 | 232 | |||
| XM_017008879.1 | XP_016864368.1 | 2 | 258 | |||
| GenBank mRNA | AB014560.1 | - | 2 | 204 | ||
| AF051311.1 | - | 1 | 84 | |||
| AF053535.1 | - | 2 | 279 | |||
| AF145284.1 | - | 2 | 279 | |||
| AK291772.1 | - | 2 | 314 | |||
| AK291786.1 | - | 2 | 219 | |||
| BC011731.2 | - | 2 | 302 | 4337457 | ||
| BI560692.1 | - | 2 | 346 | |||
| CB996973.1 | - | 2 | 234 | |||
| CD706717.1 | - | 2 | 235 | |||
| DA094091.1 | - | 2 | 249 | |||
| DA385285.1 | - | 2 | 384 | |||
| DB241424.1 | - | 2 | 493 | |||

G3BP2
Genomic Map