siRNA Details
| Gene Symbol: PSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012455.2 | NP_036587.2 | 5 | 1623 | ||
| XM_005263634.2 | XP_005263691.1 | 5 | 1688 | |||
| XM_006712392.2 | XP_006712455.1 | 5 | 1635 | |||
| XM_006712393.2 | XP_006712456.1 | 5 | 1635 | |||
| XM_006712394.2 | XP_006712457.1 | 5 | 1635 | |||
| GenBank mRNA | AB208798.1 | - | 4 | 2062 | ||
| AJ459781.1 | - | 4 | 1461 | |||
| AK023421.1 | - | 3 | 512 | |||
| AK290045.1 | - | 5 | 1633 | |||
| BC035307.1 | - | 5 | 1525 | 4339380 | ||
| BC073151.1 | - | 5 | 1585 | 5757431 | ||
| U63127.1 | - | 4 | 1503 | |||

PSD4
Genomic Map