siRNA Details
| Gene Symbol: HIGD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099668.1 | NP_001093138.1 | 4 | 1063 | ||
| NM_001099669.1 | NP_001093139.1 | 4 | 916 | |||
| NM_014056.3 | NP_054775.2 | 4 | 892 | |||
| GenBank mRNA | AF077034.1 | - | 4 | 845 | ||
| AF145385.1 | - | 4 | 831 | |||
| AK225571.1 | - | 4 | 847 | |||
| AL110233.1 | - | 4 | 862 | |||
| BC000601.1 | - | 4 | 830 | 3346355 | ||
| BC009594.1 | - | 4 | 829 | 4109190 | ||
| BC070277.1 | - | 4 | 845 | 4686431 | ||

HIGD1A
Genomic Map