siRNA Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014170.2 | NP_054889.2 | 6 | 896 | ||
| NM_138485.1 | NP_612494.1 | 5 | 797 | |||
| GenBank mRNA | AF161484.1 | - | 6 | 935 | ||
| AK093419.1 | - | 6 | 1490 | |||
| AK289611.1 | - | 5 | 759 | |||
| AK292651.1 | - | 6 | 915 | |||
| BC010137.2 | - | 4 | 632 | |||
| BC037163.1 | - | 6 | 899 | 4620206 | ||
| BX354218.2 | - | 5 | 765 | |||

GTPBP8
Genomic Map