siRNA Details
| Gene Symbol: GNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014366.4 | NP_055181.3 | 5 | 536 | ||
| NM_206825.1 | NP_996561.1 | 5 | 633 | |||
| NM_206826.1 | NP_996562.1 | 5 | 486 | |||
| GenBank mRNA | AF191018.1 | - | 5 | 533 | ||
| AK027514.1 | - | 5 | 537 | |||
| AK027516.1 | - | 5 | 386 | |||
| AK315484.1 | - | 5 | 449 | |||
| AY825265.1 | - | 4 | 363 | |||
| BC001024.2 | - | 5 | 384 | 3342771 | ||
| BF732788.1 | - | 5 | 486 | |||
| CF272475.1 | - | 5 | 405 | |||

GNL3
Genomic Map