siRNA Details
| Gene Symbol: ZBTB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316902.1 | NP_001303831.1 | 7 | 1103 | ||
| NM_001316903.1 | NP_001303832.1 | 6 | 1029 | |||
| NM_014383.2 | NP_055198.1 | 7 | 2043 | |||
| XM_011526718.2 | XP_011525020.1 | 6 | 2204 | |||
| XM_017026588.1 | XP_016882077.1 | 4 | 1817 | |||
| XM_017026589.1 | XP_016882078.1 | 7 | 2414 | |||
| XM_017026590.1 | XP_016882079.1 | 3 | 1815 | |||
| XM_017026591.1 | XP_016882080.1 | 6 | 1217 | |||
| XM_017026592.1 | XP_016882081.1 | 4 | 897 | |||
| GenBank mRNA | AF130255.1 | - | 6 | 1895 | ||
| BC017700.2 | - | 4 | 1668 | 4419776 | ||

ZBTB32
Genomic Map