siRNA Details
| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014453.3 | NP_055268.1 | 6 | 876 | ||
| NM_198426.2 | NP_940818.1 | 6 | 878 | |||
| XM_005258746.2 | XP_005258803.1 | 5 | 1505 | |||
| XM_005258747.3 | XP_005258804.1 | 5 | 1628 | |||
| GenBank mRNA | AF042384.1 | - | 6 | 844 | ||
| BC002502.2 | - | 6 | 848 | 3051024 | ||
| BU147101.1 | - | 4 | 523 | |||

CHMP2A
Genomic Map