siRNA Details
| Gene Symbol: JOSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014876.5 | NP_055691.1 | 4 | 1404 | ||
| XM_005261876.2 | XP_005261933.1 | 5 | 1711 | |||
| XM_005261877.2 | XP_005261934.1 | 5 | 1607 | |||
| XM_005261878.2 | XP_005261935.1 | 5 | 1635 | |||
| XM_005261879.2 | XP_005261936.1 | 5 | 1730 | |||
| GenBank mRNA | AK291827.1 | - | 4 | 1197 | ||
| BC015026.2 | - | 4 | 1238 | 3921785 | ||
| D31884.1 | - | 4 | 1216 | |||

JOSD1
Genomic Map