siRNA Details
| Gene Symbol: JOSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014876.5 | NP_055691.1 | 4 | 2752 | ||
| XM_005261876.2 | XP_005261933.1 | 5 | 3059 | |||
| XM_005261877.2 | XP_005261934.1 | 5 | 2955 | |||
| XM_005261878.2 | XP_005261935.1 | 5 | 2983 | |||
| XM_005261879.2 | XP_005261936.1 | 5 | 3078 | |||
| GenBank mRNA | BC015026.2 | - | 4 | 2586 | 3921785 | |
| BX538323.1 | - | 1808 | ||||
| D31884.1 | - | 4 | 2564 | |||

JOSD1
Genomic Map