siRNA Details
| Gene Symbol: TRAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265608.1 | NP_001252537.1 | 14 | 4312 | ||
| NM_001265609.1 | NP_001252538.1 | 13 | 4175 | |||
| NM_014965.4 | NP_055780.2 | 13 | 4115 | |||
| GenBank mRNA | BX647199.1 | - | 13 | 4116 | ||

TRAK1
Genomic Map