siRNA Details
| Gene Symbol: SEL1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297592.1 | NP_001284521.1 | 4 | 889 | ||
| NM_001297594.1 | NP_001284523.1 | 4 | 911 | |||
| NM_015187.4 | NP_056002.2 | 4 | 1106 | |||
| XM_011513819.2 | XP_011512121.2 | 4 | 1100 | |||
| XM_017007923.1 | XP_016863412.1 | 3 | 744 | |||
| GenBank mRNA | AB018289.1 | - | 3 | 604 | ||
| AK289687.1 | - | 4 | 854 | |||
| BC060867.1 | - | 4 | 899 | 30344157 | ||
| CR749817.1 | - | 4 | 889 | |||

SEL1L3
Genomic Map