siRNA Details
| Gene Symbol: C22orf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015370.1 | NP_056185.1 | 2 | 332 | ||
| XM_011530096.2 | XP_011528398.1 | 2 | 300 | |||
| XM_017028741.1 | XP_016884230.1 | 2 | 255 | |||
| GenBank mRNA | AL035364.1 | - | 2 | 332 | ||
| BC074758.2 | - | 2 | 250 | 30915217 | ||
| BC126262.1 | - | 2 | 254 | 8991978 | ||
| BC126264.1 | - | 2 | 254 | 8991980 | ||
| CR456405.1 | - | 2 | 219 | |||

C22orf31
Genomic Map