siRNA Details
| Gene Symbol: FAM19A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082967.2 | NP_001076436.1 | 4 | 1270 | ||
| NM_015381.6 | NP_056196.2 | 4 | 1291 | |||
| GenBank mRNA | AF131851.1 | - | 4 | 1236 | ||
| AK123443.1 | - | 5 | 1230 | |||
| AK125256.1 | - | 4 | 1291 | |||
| AY325118.1 | - | 4 | 1265 | |||
| AY358847.1 | - | 5 | 1166 | |||
| BC039396.1 | - | 4 | 1250 | 5299791 | ||

FAM19A5
Genomic Map