siRNA Details
| Gene Symbol: FAM19A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082967.2 | NP_001076436.1 | 3 | 419 | ||
| NM_015381.6 | NP_056196.2 | 3 | 440 | |||
| GenBank mRNA | AF131851.1 | - | 3 | 385 | ||
| AK123443.1 | - | 4 | 379 | |||
| AK125256.1 | - | 3 | 440 | |||
| AY325118.1 | - | 3 | 414 | |||
| AY358847.1 | - | 4 | 315 | |||
| BC039396.1 | - | 3 | 398 | 5299791 | ||

FAM19A5
Genomic Map