siRNA Details
| Gene Symbol: PARM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015393.3 | NP_056208.2 | 4 | 1747 | ||
| XM_011531833.1 | XP_011530135.1 | 5 | 1811 | |||
| GenBank mRNA | AK022311.1 | - | 4 | 1706 | ||
| AL080121.1 | - | 4 | 1663 | |||
| AY358777.1 | - | 4 | 1692 | |||

PARM1
Genomic Map