siRNA Details
| Gene Symbol: SMARCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003074.3 | NP_003065.3 | 1 | 205 | ||
| GenBank mRNA | BC021862.1 | - | 1 | 142 | ||
| BC032564.1 | - | 1 | 189 | |||
| BC039843.1 | - | 1 | 109 | |||
| BC050564.1 | - | 1 | 134 | 3912713 | ||
| BC065253.1 | - | 1 | 174 | |||
| BC113465.1 | - | 1 | 153 | 8322517 | ||
| BC117213.1 | - | 1 | 153 | 40125764 | ||
| BG474088.1 | - | 1 | 206 | |||
| U66615.1 | - | 1 | 141 | |||

SMARCC1
Genomic Map