siRNA Details
| Gene Symbol: ERLEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127397.2 | NP_001120869.1 | 13 | 2219 | ||
| NM_001127398.2 | NP_001120870.1 | 13 | 2135 | |||
| NM_015701.4 | NP_056516.2 | 14 | 2297 | |||
| GenBank mRNA | AF131743.1 | - | 6 | 1116 | ||
| AF131849.1 | - | 8 | 1486 | |||
| AY358717.1 | - | 13 | 2019 | |||
| AY453410.1 | - | 14 | 2148 | |||
| BC013129.1 | - | 14 | 2148 | 3447840 | ||
| EU794676.1 | - | 14 | 2297 | |||

ERLEC1
Genomic Map