siRNA Details
Gene Symbol: SELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016275.3 | NP_057359.2 | 6 | 1853 | ||
GenBank mRNA | AK075006.1 | - | 6 | 1826 | ||
BC026350.1 | - | 6 | 1833 | 4796883 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016275.3 | NP_057359.2 | 6 | 1853 | ||
GenBank mRNA | AK075006.1 | - | 6 | 1826 | ||
BC026350.1 | - | 6 | 1833 | 4796883 |