siRNA Details
| Gene Symbol: SNAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322902.1 | NP_001309831.1 | 4 | 416 | ||
| NM_001322903.1 | NP_001309832.1 | 5 | 512 | |||
| NM_001322904.1 | NP_001309833.1 | 5 | 482 | |||
| NM_001322905.1 | NP_001309834.1 | 5 | 445 | |||
| NM_001322906.1 | NP_001309835.1 | 5 | 439 | |||
| NM_001322907.1 | NP_001309836.1 | 6 | 640 | |||
| NM_001322908.1 | NP_001309837.1 | 6 | 563 | |||
| NM_001322909.1 | NP_001309838.1 | 5 | 430 | |||
| NM_001322910.1 | NP_001309839.1 | 5 | 537 | |||
| NM_003081.4 | NP_003072.2 | 4 | 385 | |||
| NM_130811.3 | NP_570824.1 | 4 | 385 | |||
| XM_005260808.4 | XP_005260865.1 | 4 | 463 | |||
| XM_017028021.1 | XP_016883510.1 | 5 | 315 | |||
| XM_017028022.1 | XP_016883511.1 | 5 | 407 | |||
| XM_017028023.1 | XP_016883512.1 | 4 | 489 | |||
| GenBank mRNA | AK094560.1 | - | 4 | 353 | ||
| AK098347.1 | - | 5 | 258 | |||
| AK098767.1 | - | 4 | 255 | |||
| AK223617.1 | - | 4 | 347 | |||
| AK289647.1 | - | 6 | 546 | |||
| AK314359.1 | - | 4 | 296 | |||
| BC010647.1 | - | 4 | 274 | 3867544 | ||
| BI552777.1 | - | 4 | 358 | |||
| BI602062.1 | - | 4 | 364 | |||
| BI604120.1 | - | 4 | 362 | |||
| BT019684.1 | - | 3 | 142 | |||
| BX647443.1 | - | 4 | 257 | |||
| D21267.1 | - | 4 | 347 | |||
| DA072367.1 | - | 4 | 385 | |||
| DA162693.1 | - | 4 | 489 | |||
| HY127744.1 | - | 4 | 419 | |||
| L19760.1 | - | 4 | 230 | |||
| L19761.1 | - | 4 | 230 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001291056, NM_011428, M22012, BC018249, AK078038, and others.

SNAP25
Genomic Map