siRNA Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128150.1 | NP_001121622.1 | 4 | 1191 | ||
| NM_016467.4 | NP_057551.1 | 5 | 1301 | |||
| XM_005246970.2 | XP_005247027.1 | 4 | 1220 | |||
| XM_017005347.1 | XP_016860836.1 | 5 | 1340 | |||
| GenBank mRNA | AK074756.1 | - | 4 | 1192 | ||
| AK075160.1 | - | 3 | 1194 | |||

ORMDL1
Genomic Map