siRNA Details
| Gene Symbol: ZC3H7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017590.5 | NP_060060.3 | 23 | 4823 | ||
| XM_011530016.2 | XP_011528318.1 | 23 | 4745 | |||
| GenBank mRNA | AB028954.1 | - | 20 | 4436 | ||
| AL136802.1 | - | 1 | 628 | |||
| BC047002.1 | - | 1 | 798 | |||
| BC098378.1 | - | 23 | 4761 | |||
| BC152558.1 | - | 23 | 4801 | 8860405 | ||

ZC3H7B
Genomic Map