siRNA Details
| Gene Symbol: DGKQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001347.3 | NP_001338.2 | 19, 20 | 2401 | ||
| XM_011513411.1 | XP_011511713.1 | 19, 20 | 2386 | |||
| GenBank mRNA | AB209503.1 | - | 19, 20 | 2152 | ||
| BC063801.1 | - | 19, 20 | 2409 | 6526640 | ||
| GU727647.1 | - | 19, 20 | 2339 | |||
| L38707.1 | - | 19, 20 | 2361 | |||

DGKQ
Genomic Map