siRNA Details
| Gene Symbol: MFSD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017694.3 | NP_060164.3 | 3 | 1023 | ||
| XM_005246654.2 | XP_005246711.1 | 2 | 901 | |||
| XM_005246656.3 | XP_005246713.1 | 2 | 901 | |||
| XM_011511368.1 | XP_011509670.1 | 4 | 1089 | |||
| XM_017004392.1 | XP_016859881.1 | 3 | 1023 | |||
| GenBank mRNA | AB472299.1 | - | 1 | 699 | ||
| AK098085.1 | - | 1 | 434 | |||
| AK127176.1 | - | 3 | 1023 | |||

MFSD6
Genomic Map