siRNA Details
| Gene Symbol: CC2D1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017721.4 | NP_060191.3 | 21 | 2503 | ||
| XM_005259972.2 | XP_005260029.1 | 21 | 2448 | |||
| XM_005259973.2 | XP_005260030.1 | 21 | 2451 | |||
| XM_005259974.2 | XP_005260031.1 | 21 | 2439 | |||
| XM_005259975.2 | XP_005260032.1 | 21 | 2436 | |||
| GenBank mRNA | AB097002.1 | - | 21 | 2445 | ||
| AF536205.1 | - | 23 | 2556 | |||
| AK023399.1 | - | 14 | 1328 | |||
| AK026371.1 | - | 5 | 341 | |||
| AK315068.1 | - | 21 | 2421 | |||
| BC006556.2 | - | 16 | 1607 | |||
| BC048345.1 | - | 7 | 515 | |||
| BC064981.1 | - | 21 | 2407 | 6585236 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145970, BC030439, BC027028, DQ329239, BC016188.

CC2D1A
Genomic Map