siRNA Details
| Gene Symbol: TTC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193509.1 | NP_001180438.1 | 2, 3 | 294 | ||
| NM_017735.4 | NP_060205.3 | 2, 3 | 541 | |||
| XM_005264416.1 | XP_005264473.1 | 2, 3 | 528 | |||
| XM_011532958.1 | XP_011531260.1 | 2, 3 | 528 | |||
| GenBank mRNA | AK027570.1 | - | 2, 3 | 450 | ||
| AK299196.1 | - | 2, 3 | 283 | |||
| BC001248.1 | - | 1, 2 | 74 | |||
| BC063791.1 | - | 2, 3 | 499 | 6500951 | ||

TTC27
Genomic Map