siRNA Details
| Gene Symbol: TTC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193509.1 | NP_001180438.1 | 6 | 710 | ||
| NM_017735.4 | NP_060205.3 | 6 | 957 | |||
| XM_005264416.1 | XP_005264473.1 | 6 | 944 | |||
| XM_011532958.1 | XP_011531260.1 | 6 | 944 | |||
| GenBank mRNA | AK000279.1 | - | 3 | 165 | ||
| AK027570.1 | - | 6 | 866 | |||
| AK225295.1 | - | 3 | 165 | |||
| AK299196.1 | - | 6 | 699 | |||
| BC001248.1 | - | 5 | 490 | |||
| BC063791.1 | - | 6 | 915 | 6500951 | ||

TTC27
Genomic Map