siRNA Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256766.1 | NP_001243695.1 | 2, 3 | 183 | ||
| NM_017814.2 | NP_060284.1 | 2, 3 | 183 | |||
| XM_017026916.1 | XP_016882405.1 | 2, 3 | 154 | |||
| GenBank mRNA | AK000429.1 | - | 2, 3 | 140 | ||
| AK096964.1 | - | 2, 3 | 148 | |||
| AK225424.1 | - | 2, 3 | 136 | |||
| AY358088.1 | - | 2, 3 | 167 | |||
| BC005210.1 | - | 2, 3 | 143 | 4041226 | ||
| BC016478.2 | - | 1, 2 | 107 | |||
| DA268563.1 | - | 2, 3 | 183 | |||

TMEM161A
Genomic Map