siRNA Details
| Gene Symbol: SLC25A38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017875.2 | NP_060345.2 | 6 | 1039 | ||
| XM_006713214.1 | XP_006713277.1 | 7 | 1002 | |||
| XM_011533869.2 | XP_011532171.1 | 7 | 1902 | |||
| GenBank mRNA | AK000558.1 | - | 6 | 1015 | ||
| AK026356.1 | - | 2 | 32 | |||
| AK225388.1 | - | 6 | 991 | |||
| BC013194.1 | - | 6 | 700 | 3918744 | ||
| CR457242.1 | - | 6 | 638 | |||

SLC25A38
Genomic Map